PCA - Plate Count Agar
Leverandør: DOEHLER GROUP
876631782
DOEH876631782EA 1450 NOK
DOEH876631782
PCA - Plate Count Agar
Mikrobiologiske medier Media, preparerte for mikrobiologi
Agar medium for the quantitative detection of the total germ count in the beverage and food industry.
  • Ready-to-use, liquefaction possible directly in the bottle
  • Universal applicable (membrane filtration, pouring and spread plate method)
  • Reference medium, pH neutral
The number of detectable microorganisms is an important indicator for quality assurance. The medium is also known as Bacteria Count Agar or Caseine-peptone Dextrose Yeast Agar and complies with the requirements of EN ISO 4833 and ISO 17410.