LMDA (Lee's Multi-Differential Agar)
Leverandør: DOEHLER GROUP
923555244
DOEH923555244EA 7700 NOK
DOEH923555244
LMDA (Lee's Multi-Differential Agar)
Mikrobiologiske medier Media, preparerte for mikrobiologi
Siebel-Instituts agar medium for the selective detection and quantitative determination of microorganisms (bacteria, yeasts) from the brewing environment.
  • Certified by the Siebel-Institute for technology
  • Ready-to-use powder for easy handling
  • Functionality tested with strains originally isolated from the brewing environment
Identification of the target bacteria is facilated by characteristic colour changes of the medium. In addition, acid-forming bacteria show a typical halo formation due to the breakdown of the calcium carbonate contained in the LMDA medium.