VWR® Q Path® Easyfix, Cytology Fixative
Leverandør: VWR Q-Path Chemicals
EAS0020AF59001EA 815 NOK
EAS0020AF59001 EAS0060CF59001 11056746.
VWR® Q Path® Easyfix, Cytology Fixative
Farger Microscopy and staining reagents
Cytology fixative specially designed for the conservation and transport of cytology specimens.
  • Ready-to-used pot
Suitable for laboratory use and chemical production.