HistoChoice® Clearing agent, Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
H2779-1L
H2779-1LEA 758 NOK
H2779-1L
HistoChoice® Clearing agent, Sigma-Aldrich®
Farger Microscopy and staining reagents
HistoChoice® is a patented, complex mixture of agents that allow tissues to retain their structure, antigenic sites and nucleic acid sites. This formulation in non-citrus, contains no mercury, formaldehyde or glutaraldehyde.
  • Non-toxic and odorless
  • Preserves native antigenic and nucleic acid sites
  • Tissues retain original structure
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of formalin for tissue processing.
HistoChoice® Clearing Agent is suitable as an alternative to toluene and xylene for dewaxing paraffin tissue sections. Prepared slides are suitable for antibody probing applications and in situ hybridizations.
HistoChoice is a registered trademark of Amresco, Inc.