ImmunoHistoMount™ Mounting Medium
Leverandør: SIGMA ALDRICH MICROSCOPY
I1161-30ML
I1161-30MLEA 800 NOK
I1161-30ML
ImmunoHistoMount™ Mounting Medium
Farger Microscopy and staining reagents
ImmunoHistoMount™ is a permanent aqueous mounting medium designed for tissue sections and cell smears with peroxidase and alkaline phosphatase chromogens that cannot be dehydrated with organic solvents.
  • Aqueous-based mounting medium comparable to Crystal Mount™
  • Can be used in hematology and histology applications
This mounting medium preserves fast red, aminoethylycarbazole (AEC), BCIP/NBT, and BCIP/INT chromogens and is compatible with counterstains like hematoxylin and nuclear fast red (NFR). ImmunoHistoMount is also suitable for chromogens like DAB and DAB with nickel and cobalt. It is not compatible with routine H & E staining.