ROT.ROTATEMP me-vi Viscometer Package
Leverandør: IKA
10014015
IKAA10014015EA 0 NOK
IKAA10014015
ROT.ROTATEMP me-vi Viscometer Package
Viskosimetere Rotasjonsviskosimetere
The ROTATEMP me-vi package is the ideal solution for precise and reliable viscosity measurement of difficult-to-measure, highly viscous samples.
  • Fast, precise and reproducible results: The combination of ROTAVISC viscometer and ROTATEMP high-temperature system simplifies the viscosity measurement of difficult-to-measure, highly viscous samples
  • Simple operation and space-saving: The complete system is supplied ready for connection. With its space-saving design, it fits easily into any laboratory
  • Optimum compatibility and automation: ROTAVISC and ROTATEMP
More Product Information
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Prøvevolum 600 ml
Display TFT
Driftstemperatur −100...+300 °C
B×D×H 351×372×629 mm