Open access transfer station, PuriCare®
Leverandør: LABCONCO
3820020 3820023 3820033
LBCP3820020EA 0 NOK
LBCP3820020 LBCP3820023 LBCP3820033
Open access transfer station, PuriCare®
Avtrekksskap og kabinetter Laminert flowbenk
These PuriCare Open Access Transfer Station offer utmost efficiency for cage changing operations. Open access on two sides provides ample workspace for one or two operators to transfer animals from dirty cages to clean ones. This enclosure provides personnel, animal and environmental protection. HEPA-filtered vertical airflow provides Class 100 clean air in the work area to protect the animals from cross-contamination. At the same time, room air is drawn into grilles located on the work surface directly below the sash openings to ensure containment and protect the operator from allergens.More Product Information
Bestill nå
SPESIFIKASJONER
Modell PuriCare® PuriCare® PuriCare®
Environmentally Preferable Sustainable Materials Sustainable Materials Sustainable Materials
Filtre 2 HEPA filters 99,99% efficient 2 HEPA filters 99,99% efficient 2 HEPA filters 99,99% efficient
Strømforsyning 230 V, 50 Hz, 7 A 230 V, 50/60 Hz 230 V, 50/60 Hz
Utvendig B×D×H 1483×762×1946 - 2200 mm 1006×762×1946 - 2200 mm 1232×762×1946 - 2200 mm
Innvendig B×D×H 1230×760×710 mm 1006×760×711 mm 1232×760×711 mm
Vekt 272 kg 250 kg 272 kg