Nexus™ Horizontal Clean Bench, 6' Wide
Leverandør: LABCONCO
5360630
LBCP5360630EA 0 NOK
LBCP5360630
Nexus™ Horizontal Clean Bench, 6' Wide
Avtrekksskap og kabinetter Laminert flowbenk
Nexus Horizontal Clean Benches are designed to protect non-hazardous applications, and meet the rigorous demands of life sciences, materials science applications, pharmaceutical R&D and compounding pharmacies. The Nexus also provides an ISO Class 4 clean airflow environment for protection of critical, non-hazardous samples.
  • Electronically Commutated Motor (ECM)
  • Unidirectional, laminar airflow
  • Constant Airflow Profile™ (CAP) airflow monitoring system
  • Glass side panels
  • Spill-Stopper design protects HEPA filter from spills
  • USB port for data record export