Masterflex® Transfer Tubing, Polyimide, Avantor®
Leverandør: Avantor Fluid Handling
95820-00 95820-01 95820-02 95820-03 95820-04 95820-05 95820-06 95820-07 95820-08 95820-09 95820-10 95820-11 95820-12 95820-13
MFLX95820-00EA 760 NOK
MFLX95820-00 MFLX95820-01 MFLX95820-02 MFLX95820-03 MFLX95820-04 MFLX95820-05 MFLX95820-06 MFLX95820-07 MFLX95820-08 MFLX95820-09 MFLX95820-10 MFLX95820-11 MFLX95820-12 MFLX95820-13
Masterflex® Transfer Tubing, Polyimide, Avantor®
Slanger Overføringsslange
Provides excellent temperature stability.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
The tubing is ideal for use as an electrical and thermal insulator, for fluid and gas delivery, or as a tough sheath for glass fibers. It offers excellent chemical, heat, radiation, and abrasion resistance and highMore Product Information