Microarray scanner, GenePix® 4000B
Leverandør: MOLECULAR DEVICES
GENEPIX4000B-U
MLDVGENEPIX4000B-UEA 0 NOK
MLDVGENEPIX4000B-U
Microarray scanner, GenePix® 4000B
Microarray instrumenter
The GenePix® 4000B microarray scanner is a benchmark for quality, reliability and ease-of-use in microarray scanning technology. Coupled with GenePix® Pro microarray image analysis software and Acuity® microarray informatics software, the GenePix system sets the highest standards in the acquisition and analysis of data from all types of arrays, including nucleic acids, proteins, tissues, and cells.
  • Simultaneous dual-laser scanning
  • Reliability and ease-of-use
  • Automated workflow
With automated photomultiplier (PMT) balancing, the fully integratedMore Product Information