Graphite spin columns, Pierce™
Leverandør: Thermo Fisher Scientific
88302
PIER88302EA 2325 NOK
PIER88302
Graphite spin columns, Pierce™
Filter Sentrifugefilter
Pierce™ Graphite spin columns improve phosphopeptide analysis by efficiently binding hydrophilic peptides and efficiently removing urea, salts and other contaminants before mass spectrometry analysis.
  • Convenient spin format for parallel processing of multiple samples
  • High-binding capacity with recovery of up to 100 µg of hydrophilic peptides per column
  • Porous graphite resin for cleaning up phosphopeptide samples before MS analysis
Enable fast and efficient capture, concentration, desalting and elution of hydrophilic peptides in less than ten minutes.