Pierce™ C18 Spin Columns
Leverandør: Thermo Fisher Scientific
89873
PIER89873EA 3275 NOK
PIER89873
Pierce™ C18 Spin Columns
Filter Sentrifugefilter
These columns are ready-to-use centrifuge columns of porous C18 reverse-phase resin with excellent binding and recovery characteristics for peptide sample preparation for mass spectrometry and other methods.
  • Each column contains 8 mg of C18 resin
  • Removes interfering contaminants by reducing signal suppression and improves signal-to-noise ratios and sequence coverage
  • Processing yields high-quality spectra and is effective for a variety of reverse-phase-compatible contaminates
  • Works with a wide variety of load volumes and concentrations with no need to