Centrifuge columns for affinity chromatography
Leverandør: Thermo Fisher Scientific
89868 89869 89896 89897 89898 69707
PIER89868EA 1375 NOK
PIER89868 PIER89869 PIER89896 PIER89897 PIER89898 PIER69707
Centrifuge columns for affinity chromatography
Filter Sentrifugefilter
PP column, PE filter, ~30 µm pore size. Pierce™ centrifuge columns are made from low protein binding polypropylene for compatibility with protein purification, and fit into standard centrifuge tubes for use in any centrifuge (1,5, 2, 15 and 50 ml).
Pierce centrifuge columns are convenient tools for handling 40 µl to 10 ml of an affinity support for purification. They can be used in either spin or gravity-flow format. The spin format reduces sample processing time and makes column washing and elution more efficient. For gravity-flow procedures and when slow processing time is desired forMore Product Information