Deluxe First Aid Case, ABS, Empty
Leverandør: SAFETY FIRST AID GROUP
C901GRN
SAFIC901GRNEA 2175 NOK
SAFIC901GRN
Deluxe First Aid Case, ABS, Empty
Førstehjelpsutstyr Førstehjelpssett
The Deluxe shatterproof ABS first aid case is the top range first aid storage solution. The cases are made with high quality materials to keep first aid products fresh in damp or dusty environments.
  • Ideal first aid case for any workplace
  • Keeps out damp and dust
  • Content dividers organise first aid items inside
  • High-quality rubber seal keeps contents fresh
  • Fitted with a quick-release wall bracket for ease of storage
The durable ABS plastic and rubber seal preserves the case's contents in any work environment, and the ergonomic handle