PCR plates, 96-well, elevated skirt, Axygen®
Leverandør: Corning
PCR-96-AB-C PCR-96-AB-CPBC
732-0665EA 805 NOK
732-0665 731-0692
PCR plates, 96-well, elevated skirt, Axygen®
Mikroplater PCR-plater
PP plates, semi-skirted. Axygen® 96-well elevated skirt PCR plate is designed to fit ABI Real-Time PCR thermal cyclers and sequencers. Can be sealed with 8/12 strip caps, AxyMat™ or sealing film.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified free from RNase, DNase and non pyrogenic for highest sample integrity
  • Single notch corner
  • AxyMat™ sealing mats autoclavable and reusable
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat I-RECs, ISO 14001 - Reynosa