PCR plates, 96-well
Leverandør: Brand
781350 781400 781354 781357 781373 781374 781372 781375 781376 781377 781378 781368 781371 781369 781366 781367
211-0327EA 4250 NOK
211-0327 211-0002 731-0167 731-0163 732-2933 732-2934 732-2932 732-2882 732-2935 732-2936 732-2937 732-2929 732-2931 732-2930 732-2927 732-2928
PCR plates, 96-well
Mikroplater PCR-plater
PP.
  • Ultra-thin walled design to facilitate constant, rapid and precise heat transfer
  • Smooth vessel interior minimises the binding of enzymes and nucleic acid to the walls
  • Raised well rims protect against cross-contamination and provide a tight seal in combination with matching sealing mat
  • Free from DNase, RNase and DNA
  • Autoclavable at 121 °C (2 bar), acc. DIN EN 285
96-well non skirted PCR plates are suitable for most commercially available thermal cyclers. Plates have blue alphanumeric code and cut corner marking.
96-well semi-skirted PCR platesMore Product Information