Incubators, IN/INplus (natural convection)
Leverandør: MEMMERT
IN110 IN110PLUS IN160 IN160PLUS IN260 IN260PLUS IN30 IN30PLUS IN450 IN450PLUS IN55 IN55PLUS IN75 IN750 IN750PLUS IN75PLUS IN160M IN160MPLUS IN450MPLUS IN55M IN750M IN750MPLUS IN260M IN260MPLUS IN55MPLUS IN110M IN30M IN30MPLUS IN75M IN75MPLUS IN110MPLUS IN450M
390-0644EA 29625 NOK
390-0644 390-0645 390-0646 390-0647 390-0648 390-0649 390-0650 390-0651 390-0652 390-0653 390-0654 390-0655 390-0656 390-0657 390-0658 390-0659 MEMMIN160M MEMMIN160MPLUS MEMMIN450MPLUS MEMMIN55M MEMMIN750M MEMMIN750MPLUS MEMMIN260M MEMMIN260MPLUS MEMMIN55MPLUS MEMMIN110M MEMMIN30M MEMMIN30MPLUS MEMMIN75M MEMMIN75MPLUS MEMMIN110MPLUS MEMMIN450M
Incubators, IN/INplus (natural convection)
Inkubatorer Standard Incubators
The range has natural and forced convection models with a temperature range up to 80 °C. There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application.
  • SingleDISPLAY models, ideal for standard applications:
  • ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, program time, time zones and daylight-saving time
  • One PT100 temperature sensor in a four wire circuit and Ethernet interface for exporting the protocol log
  • Double over-temperature protection including visual alarm: Electronic temperature monitoring with freely adjustable monitoring temperature and mechanical temperature limiter TB Class 1, according to DIN 12880, which switches off the heating if the maximum permitted temperature is exceeded by approx. 10 °C
All models have stainless steel inner chamber and housing, ensuring long working life and easy cleaning. These incubators have an integrated data logger and digital timer which is adjustable from 1 minute to 99 days and 23 hours. All models with TwinDISPLAY feature the intuitive control and logging software, AtmoCONTROL, which uses drag and drop symbols to input values. The software allows online monitoring of up to 32 connected appliances and can send an automatic alarm message to one or several email addresses. INplus units have a 4 hour sterilisation cycle at 160 °C. Complex ramping processes can be programmed quickly and easy for TwinDISPLAY models. The SetpointWAIT function ensures the process time does not start before the set temperature is reached. This feature can also be used with additional, freely positionable PT100 sensors, so that the process time starts when the set temperature is reached at all measuring points.
TwinDISPLAY models, ideal for complex ramping processes:
Intuitive, user friendly ovens with 2 TFT displays. ControlCOCKPIT with USB port. ControlCOCKPIT parameters as above
Two PT100 sensors in a four wire circuit for mutual monitoring, taking over functions in case of an error
The HeatBALANCE function allows application specific adjustment of heating power distribution (balance) between the upper and lower heating areas
Logged protocol data is displayed on the ControlCOCKPIT (max 10000 values correspond to approx. 1 week)
Ethernet interface for reading out the protocol log and for uploading and implementing programmes and for online logging
Multiple over-temperature protection with audible and visual alarm: Electronic temperature monitoring TWW/TWB adjustable on display (independent adjustable temperature safety device Class 3.1, which protects the samples and the incubator) and mechanical temperature limiter TB (protection Class 1) according to DIN 12880. AutoSAFETY automatically adjusts to the set value within a freely adjustable tolerance range. Setting individual MIN / MAX values for over- or under-temperature
The Im incubators, which have the same specifications as standard models, are classified as a Class I medical device in accordance with the EU directive 93/42/EEC.
INm or INmplus incubators with natural convection, can be used for the following applications: Microbiological tests, germ count determination, incubation of cell cultures, warming non-sterile cloths or blankets as well as tempering rinsing and infusion solutions.
Tilbehørs informasjon: For details of the range of shelves and grids, please visit website or contact your local Avantor sales office.
Bestillingsinformasjon: Supplied without shelves, please order required shelves, stainless steel grids or trays separately. For all chamber volumes > 53 litres, a minimum of 2 shelves or grids must be ordered in addition. Range of accessories and factory fitted options such as access ports, interior sockets, lighting and interior door are available on request. Please contact Avantor for further details prior to ordering as these cannot be retro-fitted.
NB: * RT = Ambient